MilliporeSigma
  • Juvenile growth reduces the influence of epithelial sodium channels on myogenic tone in skeletal muscle arterioles.
Juvenile growth reduces the influence of epithelial sodium channels on myogenic tone in skeletal muscle arterioles.
Clinical and experimental pharmacology & physiology (2016-08-26)
Lori S Kang, Shyama Masilamani, Matthew A Boegehold
ABSTRACT
Previous studies have documented that rapid juvenile growth is accompanied by functional changes in the arteriolar endothelium, but much less is known about functional changes in arteriolar smooth muscle over this period. In this study, we investigate the possible contribution of epithelial sodium channels (ENaC) to the myogenic behaviour of arterioles at two stages of juvenile growth. The effects of the ENaC inhibitor benzamil on different levels of myogenic tone were studied in isolated gracilis muscle arterioles from rats aged 21-28 days ("weanlings") and 42-49 days ("juveniles"). ENaC subunit expression in the arteriolar wall was also determined, and the interaction between ENaC and nitric oxide (NO) in regulating vascular tone was explored by combined use of benzamil and N
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Acetylcholine bromide, ≥99%