MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
The prognostic value of BCAR1 in patients with primary breast cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2004-09-28)
Lambert C J Dorssers, Nicolai Grebenchtchikov, Arend Brinkman, Maxime P Look, Simone P J van Broekhoven, Danielle de Jong, Harry A Peters, Henk Portengen, Marion E Meijer-van Gelder, Jan G M Klijn, Doorlene T H van Tienoven, Anneke Geurts-Moespot, Paul N Span, John A Foekens, Fred C G J Sweep
ABSTRACT
BCAR1, the human homologue of the rat p130Cas protein, was identified in a functional screen for human breast cancer cell proliferation resistant to antiestrogen drugs. Here, we study the prognostic value of quantitative BCAR1 levels in a large series of breast cancer specimens. A specific ELISA was developed to measure BCAR1 protein levels in 2593 primary breast tumor cytosols. Tumor levels of BCAR1 were correlated with relapse-free survival (RFS) and overall survival (OS) and compared with collected data on urokinase-type plasminogen activator (uPA) and plasminogen activator inhibitor 1 (PAI-1). In tumor cytosols, BCAR1 protein levels varied between 0.02 and 23 ng/mg protein. BCAR1 levels exhibited a positive correlation with steroid hormone receptor levels, age and menopausal status, and uPA and PAI-1 levels. The level of BCAR1 (continuous or categorized as low, intermediate, or high) was inversely related with RFS and OS time. Multivariate analysis showed that BCAR1 levels contributed independently to a base model containing the traditional prognostic factors for both RFS and OS (both P < 0.0001). When added together with uPA and PAI-1 in the multivariate model, BCAR1 contributed independently of PAI-1 and was favored over uPA. Interaction tests allowed for additional analyses of BCAR1 protein levels in clinically relevant subgroups stratified by nodal and menopausal status. The quantitative BCAR1 protein level represents a prognostic factor for RFS and OS in primary breast cancer, independent of the traditional prognostic factors and the other novel marker PAI-1.