MilliporeSigma
  • Microengineered Conductive Elastomeric Electrodes for Long-Term Electrophysiological Measurements with Consistent Impedance under Stretch.
Microengineered Conductive Elastomeric Electrodes for Long-Term Electrophysiological Measurements with Consistent Impedance under Stretch.
Sensors (Basel, Switzerland) (2015)
Hu D, et al.