MilliporeSigma
  • Hypochlorite scavenging activity of hydroxycinnamic acids evaluated by a rapid microplate method based on the measurement of chloramines.
Hypochlorite scavenging activity of hydroxycinnamic acids evaluated by a rapid microplate method based on the measurement of chloramines.
The Journal of Pharmacy and Pharmacology (2003)
Firuzi O