MilliporeSigma
  • Aberrant Activation of a Gastrointestinal Transcriptional Circuit in Prostate Cancer Mediates Castration Resistance.
Aberrant Activation of a Gastrointestinal Transcriptional Circuit in Prostate Cancer Mediates Castration Resistance.
Cancer cell (2017-11-21)
Shipra Shukla, Joanna Cyrta, Devan A Murphy, Edward G Walczak, Leili Ran, Praveen Agrawal, Yuanyuan Xie, Yuedan Chen, Shangqian Wang, Yu Zhan, Dan Li, Elissa W P Wong, Andrea Sboner, Himisha Beltran, Juan Miguel Mosquera, Jessica Sher, Zhen Cao, John Wongvipat, Richard P Koche, Anuradha Gopalan, Deyou Zheng, Mark A Rubin, Howard I Scher, Ping Chi, Yu Chen
ABSTRACT
Prostate cancer exhibits a lineage-specific dependence on androgen signaling. Castration resistance involves reactivation of androgen signaling or activation of alternative lineage programs to bypass androgen requirement. We describe an aberrant gastrointestinal-lineage transcriptome expressed in ∼5% of primary prostate cancer that is characterized by abbreviated response to androgen-deprivation therapy and in ∼30% of castration-resistant prostate cancer. This program is governed by a transcriptional circuit consisting of HNF4G and HNF1A. Cistrome and chromatin analyses revealed that HNF4G is a pioneer factor that generates and maintains enhancer landscape at gastrointestinal-lineage genes, independent of androgen-receptor signaling. In HNF4G/HNF1A-double-negative prostate cancer, exogenous expression of HNF4G at physiologic levels recapitulates the gastrointestinal transcriptome, chromatin landscape, and leads to relative castration resistance.
Sigma-Aldrich
Anti-HNF4G antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
MISSION® esiRNA, targeting human HNF1A
Sigma-Aldrich
Anti-AKR1C3 antibody, Mouse monoclonal, clone NP6.G6.A6, purified from hybridoma cell culture