MilliporeSigma
  • Clinical and radiological outcomes of 5-year drug-free remission-steered treatment in patients with early arthritis: IMPROVED study.
Clinical and radiological outcomes of 5-year drug-free remission-steered treatment in patients with early arthritis: IMPROVED study.
Annals of the rheumatic diseases (2017-10-04)
Gülşah Akdemir, Lotte Heimans, Sytske Anne Bergstra, Robbert J Goekoop, Maikel van Oosterhout, Johannes H L M van Groenendael, André J Peeters, Gerda M Steup-Beekman, Leroy R Lard, Peter B J de Sonnaville, Bernard A M Grillet, Tom W J Huizinga, Cornelia F Allaart
ABSTRACT
To determine the 5-year outcomes of early remission induction therapy followed by targeted treatment aimed at drug-free remission (DFR) in patients with early arthritis. In 12 hospitals, 610 patients with early (<2 years) rheumatoid arthritis (RA) or undifferentiated arthritis (UA) started on methotrexate (MTX) 25 mg/week and prednisone (60 mg/day tapered to 7.5 mg/day). Patients After 4 months, 387 patients were in early remission, 83 were randomised to arm 1 and 78 to arm 2. After 5 years, 295/610 (48%) patients were in remission, 26% in sustained DFR (SDFR) (≥1 year) (220/387 (57%) remission and 135/387 (35%) SDFR in the early remission group, 50% remission, 11% SDFR in the randomisation arms without differences between the arms). More patients with UA (37% vs 23% RA, p=0.001) and more anticitrullinated protein antibody (ACPA)-negative patients (37% vs 18% ACPA-positive, p<0.001) achieved SDFR.Overall, mean Health Assessment Questionnaire was 0.6 (0.5), and median (IQR) damage progression was 0.5 (0-2.7) Sharp/van der Heijde points, with only five patients showing progression >25 points in 5 years. Five years of DFR-steered treatment in patients with early RA resulted in almost normal functional ability without clinically relevant joint damage across treatment groups. Patients who achieved early remission had the best clinical outcomes. There were no differences between the randomisation arms. SDFR is a realistic treatment goal.
Product Description
Supelco
Diacetoxyscirpenol solution, ~100 μg/mL in acetonitrile, analytical standard