MilliporeSigma
BK
Hypertension research : official journal of the Japanese Society of Hypertension (2017-11-03)
Yanyan Zhang, Jingwen Liao, Lin Zhang, Shanshan Li, Ying Wu, Lijun Shi
ABSTRACT
Promoter methylation is a key mechanism in the epigenetic reprogramming of gene expression patterns. Here, we investigated the contribution of DNA methylation and the associated expression and function of large-conductance Ca
Sigma-Aldrich
Sodium hydrogen sulfate