MilliporeSigma
  • 5-Aza-2'-deoxycytidine reactivates gene expression via degradation of pRb pocket proteins.
5-Aza-2'-deoxycytidine reactivates gene expression via degradation of pRb pocket proteins.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2011-10-13)
Zhixing Zheng, Lian Li, Xiangyu Liu, Donglai Wang, Bo Tu, Lina Wang, Haiying Wang, Wei-Guo Zhu
ABSTRACT
Not only does 5-aza-2'-deoxycytidine (5-aza-CdR) induce the reexpression of silenced genes through the demethylation of CpG islands, but it increases the expression of unmethylated genes. However, the mechanism by which 5-aza-CdR activates the expression of genes is not completely understood. Here, we report that the pRb pocket proteins pRb, p107, and p130 were degraded in various cancer cell lines in response to 5-aza-CdR treatment, and this effect was dependent on the proteasome pathway. Mouse double minute 2 (MDM2) played a critical role in this 5-aza-CdR-induced degradation of pRb. Furthermore, PP2A phosphatase-induced MDM2 dephosphorylation at S260 was found to be essential for MDM2 binding to pRb in the presence of 5-aza-CdR. pRb degradation resulted in the significant reexpression of several genes, including methylated CDKN2A, RASFF1A, and unmethylated CDKN2D. Finally, knockdown of pRb pocket proteins by either RNAi or 5-aza-CdR treatment induced a significant decrease in the recruitment of SUV39H1 and an increase in the enrichment of KDM3B and KDM4A to histones around the promoter of RASFF1A and thus reduced H3K9 di- and trimethylation, by which RASFF1A expression is activated. Our data reveal a novel mechanism by which 5-aza-CdR induces the expression of both methylated and unmethylated genes by degrading pRb pocket proteins.
MATERIALS
Product Number
Brand
Product Description
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Phosphoserine Antibody, clone 4A4 (mouse IgG1), clone 4A4, Upstate®, from mouse
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-SUV39H1 Antibody, clone MG44, ascites fluid, clone MG44, Upstate®
Sigma-Aldrich
ChIPAb+ Dimethyl-Histone H3 (Lys9) - ChIP Validated Antibody and Primer Set, serum, from rabbit