MilliporeSigma
Smooth muscle contraction and relaxation.
Advances in physiology education (2003-11-25)
R Clinton Webb
ABSTRACT
This brief review serves as a refresher on smooth muscle physiology for those educators who teach in medical and graduate courses of physiology. Additionally, those professionals who are in need of an update on smooth muscle physiology may find this review to be useful. Smooth muscle lacks the striations characteristic of cardiac and skeletal muscle. Layers of smooth muscle cells line the walls of various organs and tubes in the body, and the contractile function of smooth muscle is not under voluntary control. Contractile activity in smooth muscle is initiated by a Ca(2+)-calmodulin interaction to stimulate phosphorylation of the light chain of myosin. Ca(2+) sensitization of the contractile proteins is signaled by the RhoA/Rho kinase pathway to inhibit the dephosphorylation of the light chain by myosin phosphatase, thereby maintaining force generation. Removal of Ca(2+) from the cytosol and stimulation of myosin phosphatase initiate the process of smooth muscle relaxation.