MilliporeSigma
  • ATF3 plays a key role in Kdo2-lipid A-induced TLR4-dependent gene expression via NF-κB activation.
ATF3 plays a key role in Kdo2-lipid A-induced TLR4-dependent gene expression via NF-κB activation.
PloS one (2010-12-15)
Eun-Young Kim, Hye Young Shin, Joo-Young Kim, Dong-Gun Kim, Yong-Min Choi, Hyuk-Kwon Kwon, Dong-Kwon Rhee, You-Sun Kim, Sangdun Choi
ABSTRACT
Activating transcription factor 3 (ATF3) is a negative regulator of proinflammatory cytokine expression in macrophages, and ATF3 deficient mice are more susceptible to endotoxic shock. This study addresses the role of ATF3 in the Kdo(2)-Lipid A-induced Toll-like receptor 4 (TLR4) signaling pathway in mouse embryonic fibroblasts (MEF). Kdo(2)-Lipid A upregulates ATF3 expression in wild type MEF cells and induces both nuclear factor kappa B (NF-κB) and c-Jun N-terminal kinase (JNK) activation via the TLR4 signaling pathway, while neither of these pathways is activated in ATF3-/- MEF cells. Interestingly, in contrast to Kdo(2)-Lipid A, the activation of both NF-κB and JNK by TNF-α was normal in ATF3-/- MEF cells. We found that several genes were dramatically upregulated in ATF3+/+ MEF cells in response to Kdo(2)-Lipid A treatment, while little difference was observed in the ATF3-/- MEF cells. However, we also found that the signal intensities of IκBζ in ATF3-/- MEF cells were substantially higher than those in wild type MEF cells upon microarray analyses, and upregulated IκBζ expression was detected in the cytosol fraction. Our findings indicate that ATF3 deficiency affects Kdo(2)-Lipid A-induced TLR4 signaling pathways in MEF cells, that it may upregulate IκBζ expression and that the high levels of IκBζ expression in ATF3-/- cells disrupts Kdo(2)-Lipid A-mediated signaling pathways.