MilliporeSigma
867210P
Avanti
SquareFluor 627/647, free acid
new
Avanti Research - A Croda Brand
Synonyme(s) :
(E)-2-(((E)-1-(5-carboxypentyl)-3,3-dimethylindolin-2-ylidene)methyl)-3-oxo-4-((1,3,3-trimethyl-3H-indol-1-ium-2-yl)methylene)cyclobut-1-en-1-olate
Se connecter pour consulter vos tarifs contractuels et ceux de votre entreprise/organisme
Le tarif et la disponibilité ne sont pas disponibles actuellement.
A propos de cet article
Formule empirique (notation de Hill) :
C33H36N2O4
Poids moléculaire :
524.65
Code UNSPSC :
12352211
Passer à
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
Source biologique
synthetic
Essai
>99% (TLC)
Forme
powder
Conditionnement
pkg of 1 × 10 mg (867210P-10MG)
Fabricant/nom de marque
Avanti Research - A Croda Brand
Fluorescence
λex 627 nm; λem 647 nm
Type de lipide
fluorescent lipids
Conditions d'expédition
dry ice
Température de stockage
−20°C
Chaîne SMILES 
C[N+]1=C(\C=C2/C([O-])=C(/C=C3/N(CCCCCC(O)=O)c4ccccc4C3(C)C)C2=O)C(C)(C)c2ccccc12
Description générale
Squaraine probes feature a distinctive squaric acid derived aromatic ring which leaves the probe vulnerable to nucleophilic attack which destroys its fluorescent ability. This limitation has historically prevented these probes from being studied as much as other probes. However, recent advancements in their synthetic design has helped address these challenges and brought them to the forefront of fluorescent imaging. Squaraines are valued for their extremely intense absorption bands, good photoconductivity, excellent photostability, and high quantum yield. 
Informations légales
Avanti Research is a trademark of Avanti Polar Lipids, LLC