Merck
Pancreatic beta-cells: Role of glycerol and aquaglyceroporin 7.
The international journal of biochemistry & cell biology (2010-11-09)
Myrna Virreira, Jason Perret, Christine Delporte
ABSTRACT
Pancreatic β-cells originate from gut endoderm during development. Pancreatic endocrine cells represent about 10% of the mature pancreatic cells, and β-cells represent the majority of endocrine cells. β-cells secrete insulin in response to elevation of nutrient concentrations. Insulin maintains glucose homeostasis by stimulating glucose uptake into muscle and adipose tissue. Aquaglyceroporin 7, permeable to water, glycerol and urea, is expressed in pancreatic β-cells and was recently described as being involved in the control of insulin secretion.
Product Description
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%