Merck
  • Bioactive components on immuno-enhancement effects in the traditional Chinese medicine Shenqi Fuzheng Injection based on relevance analysis between chemical HPLC fingerprints and in vivo biological effects.
Bioactive components on immuno-enhancement effects in the traditional Chinese medicine Shenqi Fuzheng Injection based on relevance analysis between chemical HPLC fingerprints and in vivo biological effects.
Journal of ethnopharmacology (2014-06-21)
Jinxu Wang, Xin Tong, Peibo Li, Menghua Liu, Wei Peng, Hui Cao, Weiwei Su
ABSTRACT
Shenqi Fuzheng Injection (SFI) is an injectable traditional Chinese herbal formula comprised of two Chinese herbs, Radix codonopsis and Radix astragali, which were commonly used to improve immune functions against chronic diseases in an integrative and holistic way in China and other East Asian countries for thousands of years. This present study was designed to explore the bioactive components on immuno-enhancement effects in SFI using the relevance analysis between chemical fingerprints and biological effects in vivo. According to a four-factor, nine-level uniform design, SFI samples were prepared with different proportions of the four portions separated from SFI via high speed counter current chromatography (HSCCC). SFI samples were assessed with high performance liquid chromatography (HPLC) for 23 identified components. For the immunosuppressed murine experiments, biological effects in vivo were evaluated on spleen index (E1), peripheral white blood cell counts (E2), bone marrow cell counts (E3), splenic lymphocyte proliferation (E4), splenic natural killer cell activity (E5), peritoneal macrophage phagocytosis (E6) and the amount of interleukin-2 (E7). Based on the hypothesis that biological effects in vivo varied with differences in components, multivariate relevance analysis, including gray relational analysis (GRA), multi-linear regression analysis (MLRA) and principal component analysis (PCA), were performed to evaluate the contribution of each identified component. The results indicated that the bioactive components of SFI on immuno-enhancement activities were calycosin-7-O-β-d-glucopyranoside (P9), isomucronulatol-7,2'-di-O-glucoside (P11), biochanin-7-glucoside (P12), 9,10-dimethoxypterocarpan-3-O-xylosylglucoside (P15) and astragaloside IV (P20), which might have positive effects on spleen index (E1), splenic lymphocyte proliferation (E4), splenic natural killer cell activity (E5), peritoneal macrophage phagocytosis (E6) and the amount of interleukin-2 (E7), while 5-hydroxymethyl-furaldehyde (P5) and lobetyolin (P13) might have negative effects on E1, E4, E5, E6 and E7. Finally, the bioactive HPLC fingerprint of SFI based on its bioactive components on immuno-enhancement effects was established for quality control of SFI. In summary, this study provided a perspective to explore the bioactive components in a traditional Chinese herbal formula with a series of HPLC and animal experiments, which would be helpful to improve quality control and inspire further clinical studies of traditional Chinese medicines.
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bovine IL2 / Interleukin-2 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Human IL-2 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)