Merck
  • Epithelia of the ovine and bovine forestomach express basolateral maxi-anion channels permeable to the anions of short-chain fatty acids.
Epithelia of the ovine and bovine forestomach express basolateral maxi-anion channels permeable to the anions of short-chain fatty acids.
Pflugers Archiv : European journal of physiology (2013-11-19)
Maria I Georgi, Julia Rosendahl, Franziska Ernst, Dorothee Günzel, Jörg R Aschenbach, Holger Martens, Friederike Stumpff
ABSTRACT
It has long been established that the absorption of short-chain fatty acids (SCFA) across epithelia stimulates sodium proton exchange. The apically released protons are not available as countercations for the basolateral efflux of SCFA anions and a suitable transport model is lacking. Patch clamp and microelectrode techniques were used to characterize an anion conductance expressed by cultured cells of the sheep and bovine rumen and the sheep omasum and to localize the conductance in the intact tissue. Cells were filled with a Na-gluconate solution and superfused with sodium salts of acetate, propionate, butyrate, or lactate. Reversal potential rose and whole cell current at +100 mV decreased with the size of the anion. Anion-induced currents could be blocked by diisothiocyanato-stilbene-2,2'-disulfonic acid (DIDS), NPPB (200 μmol l(-1)), or pCMB (1 mmol l(-1)). In patches of bovine ruminal cells, single channels were observed with a conductance for chloride (327 ± 11 pS), acetate (115 ± 8 pS), propionate (102 ± 10 pS), butyrate (81 ± 2 pS), and gluconate (44 ± 3 pS). Channels expressed by sheep rumen and omasum were similar. Microelectrode experiments suggest basolateral localization. In conclusion, forestomach epithelia express basolateral maxi-anion channels with a permeability sequence of chloride > acetate > propionate > butyrate. SCFA absorption may resemble functionally coupled transport of NaCl, with the Na(+)/K(+)-ATPase driving the basolateral efflux of the anion through a channel. Since protons are apically extruded, the model accurately predicts that influx of buffers with saliva is essential for the pH homeostasis of the ruminant forestomach.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Calcium chloride
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M