Merck
  • Phenylboronic acid as a multi-modal ligand for the capture of monoclonal antibodies: development and optimization of a washing step.
Phenylboronic acid as a multi-modal ligand for the capture of monoclonal antibodies: development and optimization of a washing step.
Journal of chromatography. A (2014-06-21)
Raquel dos Santos, Sara A S L Rosa, M Raquel Aires-Barros, Andres Tover, Ana M Azevedo
ABSTRACT
In this work, phenylboronic acid (PBA) was thoroughly investigated as a synthetic ligand for the purification of an immunoglobulin G (IgG) from a clarified cell supernatant from Chinese Hamster Ovary (CHO) cell cultures. In particular, the study was focused on the development of a washing step and in the optimization of the elution step using a serum containing supernatant. From the different conditions tested, best recoveries - 99% - and purifications - protein purity of 81% and a purification factor of 16 out of a maximum of 20 - were achieved using 100mM d-sorbitol in 10mM Tris-HCl as washing buffer and 0.5M d-sorbitol with 150mM NaCl in 10mM Tris-HCl as elution buffer. The purification outcome was also compared with protein A chromatography that revealed a recovery of 99%, 87% protein purity and 29 out of a maximum of 33 purification factor. Following the main purification, purified IgG was characterized in terms of isoelectric point, size and activity. In the end, a proof of concept was performed using two different mAbs from serum-free CHO cell cultures.
Product Description
Supelco
L-Proline, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
SAFC
L-Proline
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)