Merck
  • Amine-functionalized TiO₂ nanoparticles for highly selective enrichment of phosphopeptides.
Amine-functionalized TiO₂ nanoparticles for highly selective enrichment of phosphopeptides.
Talanta (2015-06-17)
Hailong Liu, Jiahong Zhou, Heyong Huang
ABSTRACT
Specific enrichment of trace phosphoproteins or phosphopeptides from complex biological samples prior to mass spectrometry analysis is of profound significance for in-depth phosphoproteomics. In this work, an amine-functionalized TiO2 nano-material with N'[3-(trimethoxysilyl)-propyl] diethylenetriamine (TPDA) modified on the surface of nanoparticle TiO2 (TiO2@TPDA) was successfully designed and applied for the enrichment of phosphopeptides. Compared with pure TiO2, the novel prepared TiO2@TPDA possessed lower Lewis acidity, enhanced hydrophilicity and stronger affinity to phosphopeptides, and its performance for selective and effective enrichment of phosphopeptide was investigated by the standard protein digests and human serum.
Sigma-Aldrich
Ultrapure Acetonitrile
SAFC
Iodoacetamide
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, suitable for electrophoresis, ≥99%
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, Molecular Biology, ≥99.5% (RT)
Supelco
DL-Dithiothreitol solution, 1 M in H2O