Merck
A new two-state polymer folding model and its application to α-helical polyalanine.
The journal of physical chemistry. B (2011-08-24)
Per Linse, Peter Palenčár, Tomáš Bleha
RÉSUMÉ
A new two-state polymer folding model is proposed, in which the folding of a stiff helical polymer is enabled by allowing for short sequences of coils connecting shorter and separated helices. The folding is driven by short-range attraction energy among stacked helices and is opposed by the free-energy cost of forming coils from helical monomers. Principal outcomes of the model are equilibrium distribution of the number of helices and their length in helical polymers. The proposed model is applied to α-helical polyalanine. The distribution of the number of α-helices as a function of number of alanine residues is fitted to the corresponding result from molecular dynamics simulation employing an all-atom potential model with very good agreement. The influence and significance of the fitting parameters and possible use of the two-state folding model are discussed.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Poly-DL-alanine, mol wt 1,000-5,000