Merck
  • Comparison of the Utility of RP-TLC Technique and Different Computational Methods to Assess the Lipophilicity of Selected Antiparasitic, Antihypertensive, and Anti-inflammatory Drugs.
Comparison of the Utility of RP-TLC Technique and Different Computational Methods to Assess the Lipophilicity of Selected Antiparasitic, Antihypertensive, and Anti-inflammatory Drugs.
Molecules (Basel, Switzerland) (2019-09-05)
Alina Pyka-Pająk, Wioletta Parys, Małgorzata Dołowy
RÉSUMÉ
The aim of this study was to assess the lipophilicity of selected antiparasitic, antihypertensive and non-steroidal anti-inflammatory drugs (NSAIDs) by means of reversed phase-thin layer chromatography (RP-TLC) as well by using Soczewiński-Wachtmeister's and J. Ościk's equations. The lipophilicity parameters of all examined compounds obtained under various chromatographic systems (i.e., methanol-water and acetone-water, respectively) and those determined on the basis of Soczewiński-Wachtmeister's and Ościk's equations (i.e., RMWS and RMWO) were compared with the theoretical ones (e.g., AlogPs, AClogP, milogP, AlogP, MlogP, XlogP2, XlogP3) and the experimental value of the partition coefficient (logPexp). It was found that the RMWS parameter may be a good alternative tool in describing the lipophilic nature of biologically active compounds with a high and low lipophilicity (i.e., antihypertensive and antiparasitic drugs). Meanwhile, the RMWO was more suitable for compounds with a medium lipophilicity (i.e., non-steroidal anti-inflammatory drugs). The chromatographic parameter 0(a) can be helpful for the prediction of partition coefficients, i.e., AClogP, XlogP3, as well as logPexp of examined compounds.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Supelco
Ornidazole, analytical standard