Merck
Ultraviolet photodetector with high internal gain enhanced by TiO₂/SrTiO₃ heterojunction.
Optics express (2012-03-16)
Min Zhang, Haifeng Zhang, Kaibo Lv, Weiyou Chen, Jingran Zhou, Liang Shen, Shengping Ruan
RÉSUMÉ
In this letter, TiO₂ nanocrystalline film was prepared on SrTiO₃ (001) substrate to form an n-n heterojunction active layer. Interdigitated Au electrodes were deposited on the top of TiO₂ film to fabricate modified HMSM (heterojunction metal-semiconductor-metal) ultraviolet photodetector. At 10 V bias, the dark current of the detector was only 0.2 nA and the responsivity was 46.1 A/W at 260 nm. The rise and fall times of the device were 3.5 ms and 1.4 s, respectively. The TiO₂/SrTiO₃ heterojunction contributed a lot to the high responsivity and reduced the fall time, which improved the device performance effectively. These results demonstrate the excellent application of TiO₂/SrTiO₃ heterojunction in fabricating high performance UV photodetectors.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Strontium titanate, powder, 99%
Sigma-Aldrich
Strontium titanate, nanopowder, <100 nm particle size, 99% trace metals basis
Sigma-Aldrich
Strontium titanate, single crystal substrate, <100>