Analysis Note
Density (d 20 °C/ 4 °C): 0.869 - 0.874
Assay (acidimetric): ≥ 98.0 %
Iodine value: 75 - 84
Identity (IR): passes test
Assay (acidimetric): ≥ 98.0 %
Iodine value: 75 - 84
Identity (IR): passes test
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
335.3 °F - Pensky-Martens closed cup
flash_point_c
168.5 °C - Pensky-Martens closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service