O této položce
NACRES:
NB.79
UNSPSC Code:
41116129
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohliQuality Segment
material
316 stainless steel , polyphenylsulfone (PPSU)
feature
autoclavable
manufacturer/tradename
Milliflex®
application(s)
bioburden testing
pharmaceutical
water monitoring
compatibility
for use with MILLIFLEX®, for use with Milliflex and Milliflex Oasis devices and media types
General description
Milliflex® Quantum membrane removal tool is an additional accessory for Milliflex® Quantum Rapid Detection System. It can be used with the Milliflex® system and with the Milliflex Oasis® system.Milliflex® Quantum System is a rapid fluorescence-based technology for fast quantitative detection of contaminants over a broad range of filterable matrices. This easy to use and simple system can detect viable microorganisms filtered on a membrane down to one CFU per sample. The Milliflex® Quantum system uses proven technology based on a universal enzymatic fluorescent staining of viable microorganisms. The Milliflex® Quantum membrane removal tool should be used in a microbiological safety cabinet or laminar flow hood.
Application
Milliflex® Quantum membrane removal tool is an additional accessory for Milliflex® Quantum Rapid Detection System to ensure safety removal of the membrane. It finds application in bioburden testing and water monitoring of beer, bottled water, cosmetics and pharmaceutical products analysis.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?