O této položce
Empirický vzorec (Hillův zápis):
C10H12N2O4
Číslo CAS:
Molekulová hmotnost:
224.21
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Assay:
≥97% (TLC)
Form:
powder
Solubility:
methanol: 50 mg/mL, clear, colorless to faintly yellow
Storage temp.:
−20°C
assay
≥97% (TLC)
form
powder
solubility
methanol: 50 mg/mL, clear, colorless to faintly yellow
storage temp.
−20°C
SMILES string
CC1=C[N@@H]2[C@H]3C[C@@H](OC2=NC1=O)[C@@H](CO)O3
InChI
1S/C10H12N2O4/c1-5-3-12-8-2-6(7(4-13)15-8)16-10(12)11-9(5)14/h3,6-8,13H,2,4H2,1H3/t6-,7-,8-/m1/s1
InChI key
JCSNHEYOIASGKU-BWZBUEFSSA-N
Application
2,3′-Anhydrothymidine may be derivitized to 5′-benzoyl-2′,3′-anhydrothymidine for use in the synthesis of 5′-Derivatives of 3′-(tetrazole-2′-yl)-3′-deoxythymidines. 2,3′-Anhydrothymidine may be contacted with thioacetic acid to produce 3′-S-acetyl-3′-thio-2′-deoxynucleosides.
Skladovací třída
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?