O této položce
NACRES:
NB.12
UNSPSC Code:
41122804
eCl@ss:
32032201
Manufacturer/tradename:
Heathrow Scientific HS120115
Feature:
Holds 240 x 10 mL tubes
Material:
polypropylene
Packaging:
pack of 1 ea
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
polypropylene
feature
Holds 240 x 10 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120115
size
673x267x90 mm
color
blue
General description
Mega Rack High-capacity storage rack for sample and blood collection tubes.
- Fully-assembled, polypropylene Mega Tube Racks are great for high throughput labs requiring large sample storage.
- Sturdy and light weight material for easy transport
- Alphanumeric identification for quick location and retrieval of tubes
- Place different test groupings into one rack and eliminate need for multiple tube racks
- Autoclavable
Legal Information
Mega Rack is a registered trademark of Heathrow Scientific
Still not finding the right product?
Explore all of our products under Mega Rack® tube rack
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?