Skip to Content
Merck

661880

4-Bromo-2-methoxybenzaldehyde

97%

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C8H7BrO2
CAS Number:
Molecular Weight:
215.04
UNSPSC Code:
12352100
PubChem Substance ID:
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

4-Bromo-2-methoxybenzaldehyde, 97%

InChI

1S/C8H7BrO2/c1-11-8-4-7(9)3-2-6(8)5-10/h2-5H,1H3

SMILES string

COc1cc(Br)ccc1C=O

InChI key

YMXQYZABFWVXEK-UHFFFAOYSA-N

assay

97%

form

solid

mp

67-71 °C

functional group

aldehyde
bromo

Application

Building block in the synthesis of a nonacid degradable linker for solid-phase synthesis.[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aina Colombo et al.
Organic letters, 9(21), 4319-4322 (2007-09-18)
Synthesis and applications of two new nonacid degradable linkers as an alternative to the Wang linker for solid-phase synthesis are described. Resin from linker 2 looks superior to linker 1 in terms of yields for both anchoring of the first

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service