Skip to Content
Merck

GF19068037

Zinc

foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%

Synonym(s):

Zinc, ZN000305

Sign In to View Organizational & Contract Pricing

Select a Size

10 EA

CZK 26,900.00

CZK 26,900.00


Please contact Customer Service for Availability

Request a Bulk Order

About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

vapor pressure

1 mmHg ( 487 °C)

Assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 190-680-37

resistivity

5.8 μΩ-cm, 20°C

diam. × thickness

8 mm × 0.25 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF25289837GF44141712GF84072540
Zinc foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%

GF19068037

Zinc

Zinc foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%

GF25289837

Zinc

Zinc foil, 8mm disks, thickness 0.5mm, as rolled, 99.99+%

GF44141712

Zinc

Zinc foil, 8mm disks, thickness 0.25mm, as rolled, 99.95+%

GF84072540

Zinc

assay

99.99%

assay

99.99%

assay

99.99%

assay

99.95%

manufacturer/tradename

Goodfellow 190-680-37

manufacturer/tradename

Goodfellow 252-898-37

manufacturer/tradename

Goodfellow 441-417-12

manufacturer/tradename

Goodfellow 840-725-40

form

foil

form

foil

form

foil

form

foil

mp

420 °C (lit.)

mp

420 °C (lit.)

mp

420 °C (lit.)

mp

420 °C (lit.)

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

density

7.133 g/mL at 25 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhonghao Li et al.
Chemical communications (Cambridge, England), (41)(41), 6273-6275 (2009-10-15)
Aligned ZnO arrays with branched rods, hexagonal rods, quasi-round rods, hexagonal pyramids and porous nanostructures are constructed on zinc foil in the ionic liquid tetrabutylammonium hydroxide aqueous solution.
Tandra Ghoshal et al.
Nanotechnology, 19(6), 065606-065606 (2008-02-13)
ZnO nanowire arrays were synthesized on zinc foil by a simple thermal evaporation process at relatively low temperature. Morphology and size controlled synthesis of the ZnO nanostructures was achieved by variation of the synthesis temperature, reaction time and the surface
Tandra Ghoshal et al.
Journal of nanoscience and nanotechnology, 9(9), 5586-5591 (2009-11-26)
Nanowire arrays of zinc oxide were synthesized on zinc foil by a simple thermal evaporation process. Morphologies and sizes of the synthesized nanostructures were varied by varying the reaction time and the surface roughness of the substrate. Self-catalytic liquid-solid mechanism
S Rondot et al.
Science (New York, N.Y.), 263(5154), 1739-1741 (1994-03-25)
An x-ray projection microscope equipped with a charge-coupled device camera allows direct observation of zinc (Zn(2+)) ions diffusing in aqueous hydrochloric acid solution during the corrosion of zinc foil and pellets. Time series of microradiographic images with a lateral resolution
Xufeng Wu et al.
Chemical communications (Cambridge, England), (15)(15), 1655-1657 (2006-04-04)
Highly oriented ZnO nanoneedle/nanorods arrays have been fabricated by direct oxidation of zinc foil in alkaline zincate ion solution at near room temperature (20 degrees C for nanoneedles, 30 degrees C for nanorods).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service