Skip to Content
Merck

GF87603891

Palladium

foil, 10x10mm, thickness 0.50mm, as rolled, 99.95%

Synonym(s):

Palladium, PD000350

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141733
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Palladium, foil, 10x10mm, thickness 0.50mm, as rolled, 99.95%

InChI

1S/Pd

SMILES string

[Pd]

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

assay

≥99.95%

form

foil

manufacturer/tradename

Goodfellow 876-038-91

resistivity

9.96 μΩ-cm, 20°C

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF35373551GF51548418GF95863821
manufacturer/tradename

Goodfellow 876-038-91

manufacturer/tradename

Goodfellow 353-735-51

manufacturer/tradename

Goodfellow 515-484-18

manufacturer/tradename

Goodfellow 958-638-21

form

foil

form

foil

form

foil

form

foil

assay

≥99.95%

assay

99.99%

assay

99.95%

assay

≥99.95%

mp

1554 °C (lit.)

mp

1554 °C (lit.)

mp

1554 °C (lit.)

mp

1554 °C (lit.)

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

bp

2970 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pazhamalai Anbarasan et al.
Chemical Society reviews, 40(10), 5049-5067 (2011-04-30)
The palladium-catalyzed cyanation of Ar-X (X = I, Br, Cl, OTf, and H) allows for an efficient access towards benzonitriles. After its discovery in 1973 and following significant improvements in recent decades, this methodology has become nowadays the most popular
Palladium(II)-catalyzed alkene functionalization via nucleopalladation: stereochemical pathways and enantioselective catalytic applications.
Richard I McDonald et al.
Chemical reviews, 111(4), 2981-3019 (2011-03-25)
Xiao-Feng Wu et al.
ChemSusChem, 6(2), 229-241 (2013-01-12)
Palladium-catalyzed coupling reactions have become a powerful tool for advanced organic synthesis. This type of reaction is of significant value for the preparation of pharmaceuticals, agrochemicals, as well as advanced materials. Both, academic as well as industrial laboratories continuously investigate
Qing-An Chen et al.
Chemical Society reviews, 42(2), 497-511 (2012-11-10)
The transition metal catalyzed asymmetric hydrogenation of unsaturated compounds arguably presents one of the most attractive methods for the synthesis of chiral compounds. Over the last few decades, Pd has gradually grown up as a new and popular metal catalyst
Abdellatif Essoumhi et al.
Journal of nanoscience and nanotechnology, 14(2), 2012-2023 (2014-04-23)
This review reports on the recent advances in the synthesis and physico-chemical properties of palladium-containing perovskites. Initially, the perovskite structure is briefly reviewed, then palladium-containing perovskites synthesis and physico-chemical properties are detailed. The applications of palladium-containing perovskites in catalysis; namely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service