Sign In to View Organizational & Contract Pricing
Select a Size
700394M-1MG
CZK 9,150.00
CZK 9,150.00
Please contact Customer Service for Availability
About This Item
Empirical Formula (Hill Notation):
C20H21D11O3
Molecular Weight:
331.53
UNSPSC Code:
12352211
Skip To
biological source
synthetic
Assay
>99% (TLC)
form
(Dichloromethane Solution)
packaging
pkg of 1 × 1 mg (700394M-1MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
shipped in
dry ice
storage temp.
-65 to -96°C
SMILES string
[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[C@H](O)\C=C\C=C/C\C=C/C\C=C/CCCC(O)=O
Related Categories
General description
15(S)-HETE-d11 is part of a group of acids called Hydroxyeicosatetraenoic acids (HETEs). This group of acids are metabolites of arachidonate acid and play a significant role in various tissue and organs. HETEs have been shown to play a role in the modulation of inflammation in the early stages of diabetes. 15(S)-HETE-d11 is the deuterated version which will enable the study of structure, dynamics, and function of this lipid within the cell.
Application
Lipidomics
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service