Skip to Content
Merck

880183P

Avanti

Cyanine 7, free acid

new

Avanti Research - A Croda Brand

Synonym(s):

1-(5-carboxypentyl)-3,3-dimethyl-2-((1E,3E,5E)-7-((E)-1,3,3-trimethylindolin-2-ylidene)hepta-1,3,5-trien-1-yl)-3H-indol-1-ium

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C34H41BrN2O2
Molecular Weight:
589.61
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

Assay

≥99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

fluorescent lipids

storage temp.

-10 to -25°C

SMILES string

[Br-].CN1\C(=C\C=C\C=C\C=C\C2=[N+](CCCCCC(O)=O)c3ccccc3C2(C)C)C(C)(C)c2ccccc12

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service