Skip to Content
Merck

BDPH9BCCDDCD

Bide Pharmatech

Decyl decanoate

Synonym(s):

decyl decanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H40O2
CAS Number:
Molecular Weight:
312.54
MDL number:
Product BDPH9BCCDDCD is not currently sold in your country.


Inchi Code

1S/C20H40O2/c1-3-5-7-9-11-13-15-17-19-22-20(21)18-16-14-12-10-8-6-4-2/h3-19H2,1-2H3

Inchi Key

XAKXZZPEUKNHMA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless to Almost colorless liquid



Still not finding the right product?

Explore all of our products under Decyl decanoate



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library