Skip to Content
Merck

BDPH9BCCFF4E

Bide Pharmatech

5-Bromo-3-isopropyl-1H-indole

Synonym(s):

5-bromo-3-isopropyl-1H-indole

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCCFF4E is not currently sold in your country.

About This Item

Linear Formula:
C11H12N1Br1
CAS Number:
Molecular Weight:
238.13
MDL number:
Product BDPH9BCCFF4E is not currently sold in your country.

Inchi Code

1S/C11H12BrN/c1-7(2)10-6-13-11-4-3-8(12)5-9(10)11/h3-7,13H,1-2H3

Inchi Key

FBNRLQRIMLEPOY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library