Product BDPH9BCD1D64 is not currently sold in your country.
Inchi Code
1S/C11H12O3/c1-11(2)6-8-7(10(12)13)4-3-5-9(8)14-11/h3-5H,6H2,1-2H3,(H,12,13)
Inchi Key
NVTPSSIGWHVYGN-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Country of Origin
CN
Physical Form
Solid
Storage temp.
Sealed in dry, 2-8°C
Still not finding the right product?
Explore all of our products under 2,2-Dimethyl-2,3-dihydrobenzofuran-4-carboxylic acid
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
