Product BDPH9BCD37CD is not currently sold in your country.
Inchi Code
1S/C21H30N2O11/c24-17-1-2-18(25)22(17)6-8-30-10-12-32-14-16-33-15-13-31-11-9-29-7-5-21(28)34-23-19(26)3-4-20(23)27/h1-2H,3-16H2
Inchi Key
PDFXZGGPCFZXJU-UHFFFAOYSA-N
Shipping Temperature
Cold-chain transportation
Purity
95%
Storage temp.
Keep in dark place, inert atmosphere, store in freezer, under -20C
Country of Origin
CN
Physical Form
Colorless to White to Yellow Liquid or Semi-solid or Solid
Still not finding the right product?
Explore all of our products under Mal-peg5-nhs ester
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
