Skip to Content
Merck

BDPH9BCD4BE2

Bide Pharmatech

Methyl 2-bromo-5-methylthiophene-3-carboxylate

Synonym(s):

methyl 2-bromo-5-methyl-3-thiophenecarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7O2Br1S1
CAS Number:
Molecular Weight:
235.1
MDL number:
Product BDPH9BCD4BE2 is not currently sold in your country.


Inchi Code

1S/C7H7BrO2S/c1-4-3-5(6(8)11-4)7(9)10-2/h3H,1-2H3

Inchi Key

ROKSORSRSUAPHR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, 2-8°C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library