Skip to Content
Merck

BDPH9BCD4E2F

Bide Pharmatech

(4-Amino-2-methyl-5-pyrimidinyl)methanol

Synonym(s):

(4-amino-2-methyl-5-pyrimidinyl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCD4E2F is not currently sold in your country.

About This Item

Linear Formula:
C6H9O1N3
CAS Number:
Molecular Weight:
139.16
MDL number:
Product BDPH9BCD4E2F is not currently sold in your country.

Inchi Code

1S/C6H9N3O/c1-4-8-2-5(3-10)6(7)9-4/h2,10H,3H2,1H3,(H2,7,8,9)

Inchi Key

VUTBELPREDJDDH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White - Yellow

Looking for similar products? Visit Product Comparison Guide

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library