Skip to Content
Merck

BDPH9BCD4E9E

Bide Pharmatech

6-Iodouracil

Synonym(s):

6-iodo-2,4(1H,3H)-pyrimidinedione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H3O2N2I1
CAS Number:
Molecular Weight:
237.98
MDL number:
Product BDPH9BCD4E9E is not currently sold in your country.


Inchi Code

1S/C4H3IN2O2/c5-2-1-3(8)7-4(9)6-2/h1H,(H2,6,7,8,9)

Inchi Key

KSLZHGYKGYWHOO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Keep in dark place, inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under 6-Iodouracil


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library