Skip to Content
Merck

BDPH9BCD4EEC

Bide Pharmatech

5,5-Dimethyl-2-(4-(trifluoromethyl)phenyl)-1,3,2-dioxaborinane

Synonym(s):

5,5-dimethyl-2-(4-(trifluoromethyl)phenyl)-1,3,2-dioxaborinane

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCD4EEC is not currently sold in your country.

About This Item

Linear Formula:
C12H14O2F3B1
CAS Number:
Molecular Weight:
258.05
MDL number:
Product BDPH9BCD4EEC is not currently sold in your country.

Inchi Code

1S/C12H14BF3O2/c1-11(2)7-17-13(18-8-11)10-5-3-9(4-6-10)12(14,15)16/h3-6H,7-8H2,1-2H3

Inchi Key

RHRLRPXJTDDILM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library