Product BDPH9BCD4F2E is not currently sold in your country.
Inchi Code
1S/C8H19NO2/c1-4-9-7-8(10-5-2)11-6-3/h8-9H,4-7H2,1-3H3
Inchi Key
YSCNYKGRVUVJHA-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Keep in dark place, sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under 2,2-Diethoxy-N-ethylethanamine
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

