Product BDPH9BCD652B is not currently sold in your country.
Inchi Code
1S/C8H7NO/c1-6-2-3-7(5-9)8(10)4-6/h2-4,10H,1H3
Inchi Key
WAFLXCCXXVMUKI-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Pale-yellow to Yellow-brown to Brown Solid
Still not finding the right product?
Explore all of our products under 2-Hydroxy-4-methylbenzonitrile
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
