Skip to Content
Merck

BDPH9BCD7331

Bide Pharmatech

(2-Bromothiazol-5-yl)methanamine

Synonym(s):

(2-bromo-1,3-thiazol-5-yl)methanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H5N2Br1S1
CAS Number:
Molecular Weight:
193.07
MDL number:
Product BDPH9BCD7331 is not currently sold in your country.


Inchi Code

1S/C4H5BrN2S/c5-4-7-2-3(1-6)8-4/h2H,1,6H2

Inchi Key

JZLXHPPZGZESTM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid



Still not finding the right product?

Explore all of our products under (2-Bromothiazol-5-yl)methanamine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library