Skip to Content
Merck

BDPH9BCD8E8A

Bide Pharmatech

(2-(Isopropylthio)pyrimidin-5-yl)boronic acid

Synonym(s):

(2-(isopropylthio)pyrimidin-5-yl)boronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H11O2N2S1B1
CAS Number:
Molecular Weight:
198.05
MDL number:
Product BDPH9BCD8E8A is not currently sold in your country.


Physical Form

Powder or crystals

Inchi Code

1S/C7H11BN2O2S/c1-5(2)13-7-9-3-6(4-10-7)8(11)12/h3-5,11-12H,1-2H3

Inchi Key

MEFOGGSKEYACMW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library