Product BDPH9BCDAEE8 is not currently sold in your country.
Inchi Code
1S/C6H6N2/c1-2-4-8-5-3-7-6-8/h1,3,5-6H,4H2
Inchi Key
UGOQKSGJGCADAG-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under 1-(Prop-2-yn-1-yl)-1H-imidazole
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


