Sign In to View Organizational & Contract Pricing.
Select a Size
Product BDPH9BCDB07F is not currently sold in your country.
About This Item
Product BDPH9BCDB07F is not currently sold in your country.
Inchi Code
1S/C16H12N2/c17-13-5-9-1-2-10-6-14(18)8-12-4-3-11(7-13)15(9)16(10)12/h1-8H,17-18H2
Inchi Key
JSTHREDTMPIBEX-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Looking for similar products? Visit Product Comparison Guide
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
