Product BDPH9BCDC032 is not currently sold in your country.
Inchi Code
1S/C45H31O4P/c46-50(47)48-39-19-9-18-36-38(42-33-16-7-3-12-29(33)25-30-13-4-8-17-34(30)42)26-45(43(36)39)23-22-37-35(20-21-40(49-50)44(37)45)41-31-14-5-1-10-27(31)24-28-11-2-6-15-32(28)41/h1-21,24-25,38H,22-23,26H2,(H,46,47)
Inchi Key
XQYKGXZHTZQIGY-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Powder or crystal
Still not finding the right product?
Explore all of our products under (11aS)-3,7-Di-9-anthracenyl-10,11,12,13-tetrahydro-5-hydroxy-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin-5-oxide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
