Skip to Content
Merck

BDPH9BCDCE99

Bide Pharmatech

1-(4-Bromobutyl)-4-vinylbenzene

Synonym(s):

1-(4-bromobutyl)-4-vinylbenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCDCE99 is not currently sold in your country.

About This Item

Linear Formula:
C12H15Br1
CAS Number:
Molecular Weight:
239.16
MDL number:
Product BDPH9BCDCE99 is not currently sold in your country.

Inchi Code

1S/C12H15Br/c1-2-11-6-8-12(9-7-11)5-3-4-10-13/h2,6-9H,1,3-5,10H2

Inchi Key

UCDGLMPOKKXSDM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library