Product BDPH9BCDEB09 is not currently sold in your country.
Inchi Code
1S/C9H11N.ClH/c10-1-9-6-3-2-4(6)8(9)5(2)7(3)9;/h2-8H,1,10H2;1H
Inchi Key
NVPRPIREAYXWHV-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under Cuban-1-ylmethanamine hydrochloride
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
