Skip to Content
Merck

BDPH9BCDFD42

Bide Pharmatech

Diisopropyl Sulfate

Synonym(s):

diisopropyl sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCDFD42 is not currently sold in your country.

About This Item

Linear Formula:
C6H14O4S1
CAS Number:
Molecular Weight:
182.24
MDL number:
Product BDPH9BCDFD42 is not currently sold in your country.

Inchi Code

1S/C6H14O4S/c1-5(2)9-11(7,8)10-6(3)4/h5-6H,1-4H3

Inchi Key

HWBLTYHIEYOAOL-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

97%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

colorless to light yellow liquid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library