Skip to Content
Merck

BDPH9BCDFD51

Bide Pharmatech

2,4,6-Trimethyliodobenzene

Synonym(s):

2-iodo-1,3,5-trimethylbenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11I1
CAS Number:
Molecular Weight:
246.09
MDL number:
Product BDPH9BCDFD51 is not currently sold in your country.


Inchi Code

1S/C9H11I/c1-6-4-7(2)9(10)8(3)5-6/h4-5H,1-3H3

Inchi Key

GTPNXFKONRIHRW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or Semi-solid or Liquid or Cloudy liquid or Lump



Still not finding the right product?

Explore all of our products under 2,4,6-Trimethyliodobenzene


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library