Skip to Content
Merck

BDPH9BCDFD53

Bide Pharmatech

Methyl 5-bromo-6-fluoro-1H-indole-3-carboxylate

Synonym(s):

methyl 5-bromo-6-fluoro-1H-indole-3-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9BCDFD53 is not currently sold in your country.

About This Item

Linear Formula:
C10H7O2N1Br1F1
CAS Number:
Molecular Weight:
272.07
MDL number:
Product BDPH9BCDFD53 is not currently sold in your country.

Inchi Code

1S/C10H7BrFNO2/c1-15-10(14)6-4-13-9-3-8(12)7(11)2-5(6)9/h2-4,13H,1H3

Inchi Key

WNEWBCYCMPFEBA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library