Skip to Content
Merck

BDPH9BCDFF8B

Bide Pharmatech

1-(Difluoromethyl)-1H-imidazole-2-carbaldehyde

Synonym(s):

1-(difluoromethyl)-1H-imidazole-2-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H4O1N2F2
CAS Number:
Molecular Weight:
146.1
MDL number:
Product BDPH9BCDFF8B is not currently sold in your country.


Inchi Code

1S/C5H4F2N2O/c6-5(7)9-2-1-8-4(9)3-10/h1-3,5H

Inchi Key

PPQLGRBFICLBCJ-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid or semi-solid or lump or liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library