Skip to Content
Merck

BDPH9C021BBB

Bide Pharmatech

N-[(1S,2S)-2-Amino-1,2-diphenylethyl]-1,1,1-trifluoromethanesulfonamide

Synonym(s):

N-((1S,2S)-2-amino-1,2-diphenylethyl)-1,1,1-trifluoromethanesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9C021BBB is not currently sold in your country.

About This Item

Linear Formula:
C15H15O2N2S1F3
CAS Number:
Molecular Weight:
344.36
MDL number:
Product BDPH9C021BBB is not currently sold in your country.

Inchi Code

1S/C15H15F3N2O2S/c16-15(17,18)23(21,22)20-14(12-9-5-2-6-10-12)13(19)11-7-3-1-4-8-11/h1-10,13-14,20H,19H2/t13-,14-/m0/s1

Inchi Key

GIBMTDKHCXCSNA-KBPBESRZSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to off-white powder or crystals

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library